Learn More
Bel-Art™ 96 Deep-Well Plates, Automation Compliant
Conforms to SBS/ANSI/SLAS footprints
Brand: Bel-Art™ F37860-0007
51.95 EUR valid until 2025-12-31
Use promo code "24288" to get your promotional price.
This item is not returnable.
View return policy
Description
- Used with multi-channel pipettes or with automation systems
- Made of virgin polypropylene
- DNase, RNase, pyrogen free
- Manufactured in Class 8 clean room
- Alphanumeric marking and cut-angle mark for easy referencing
- Can be stored at -80 °C
- Autoclavable up to 121 °C
- CE marked
Types:
- V-bottom shape
- For ease of sample retrieval
- U-bottom shape
- Ideal for agglutination or applications where light stirring is necessary
Specifications
No Barcode | |
Standard | |
12 x 8 | |
Transparent | |
Multi-channel pipettes or automation systems | |
2.2 mL | |
Non-sterile | |
2.2 mL | |
SBS/ANSI/SLAS footprints |
Automation compliant, CE marked | |
Store at -80 °C | |
Autoclavable | |
Well Plate | |
Polypropylene | |
96 | |
Non-treated | |
Conical Bottom (V-well) | |
5 Pk. |