Learn More
Brady™ All-weather Vinyl Labels
Make labels that withstand up to 10 years of outdoor abuse with this low-shrink vinyl label material (B-595). Print almost anywhere with a durable Brady™ handheld label maker.
Brand: Brady™ 170942
This item is not returnable.
View return policy
Description
- Ideal for labeling pipes, equipment, shelves and other industrial surfaces
- Designed to withstand prolonged exposure to UV light, chemicals, greases and oils, salt spray, cleaners and detergents, and grime
- Made with high-tack, permanent acrylic adhesive for use on rough, highly-textured, dirty and smooth surfaces alike, especially in cold temperatures
- Weather-resistant labels designed to last up to 10 years in an outdoor environment
Specifications
Not Autoclavable | |
Permanent Acrylic | |
Orange | |
Continuous | |
Top | |
Labels | |
Component and equipment labelling;Electrical labelling;Facility identification;Floor marking;Lean and 5S labelling;Pipe and valve marking;Warehouse marking | |
-40°C | |
1 Label(s) / Box | |
38.10 mm (W) x 7.62 m (L) | |
-40°C to 82°C | |
0.102 mm | |
25 ft. | |
1.5 in | |
38.10 mm | |
M5C-1500-595-OR-BK |
BS 5609:1986 Section 2 and section 3 Compliant with Directive 2011/65/EU and 2015/863 | |
None | |
Gloss | |
Vinyl | |
Thermal Transfer | |
Non-sterile | |
BMP51, BMP53, M511, M510 | |
M5C-1500-595-OR-BK | |
1 | |
-40°F to 180°F | |
0.004 in | |
38.10 mm (W) x 7.62 m (L) | |
7.62 m | |
38.10 mm | |
0.5 in. |