Gibco™ Fetal Bovine Serum, Premium Plus
Gibco Premium Plus, our most defined FBS provides consistent cell growth for sensitive cell lines requiring the lowest viral risk. Premium Plus is ideal for use in biomanufacturing processes and diagnostic cell culture research requiring the most stringent quality profile, including an extensive biochemical and hormonal profile. We offer a variety of Sera including Gamma Irradiated and Heat-Inactivated.
Overview
101.65 EUR valid until 2025-12-31
Use promo code "24268" to get your promotional price.
Additional Details:
Additional Details: Weight: 0.04540kg
Disclaimers: For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Product Code. 17964721
Quantity Price
1 110.0  € / 50mL
In stock, ready to ship: 24
Add to basket
Description and Specification
Specification
Content And Storage Store at ≤-10°C.
Species Cattle/Bovine
Age Fetal
Country of Origin Refer to CoA for specific origin
Form Frozen
Format One Shot
Product Type Fetal Bovine Serum
Quantity 50 mL
Serum Treatment Standard (Sterile-filtered)
Sterility Sterile
Sterilization Method Triple-filtered, 0.1 μm
Grade Premium Plus
Processing Triple-filtered, 0.1 μm
Tested For Up to 70 quality tests including EMA virus testing; USP/EP mycoplasma, endotoxin, performance; biochemical/hormonal profiling; Oritain fingerprinting
Endotoxin Concentration ≤5 EU/mL
Green Features Less waste
Heat Inactivated Yes
Hemoglobin Concentration ≤20 mg/dL
Shipping Condition Dry Ice
USDA Safety Testing Included USDA Safety Testing Performed for Australian origins
Gammairradiated No
Sera category: Premium Plus
  • Use for a broad range of cell types, especially sensitive cell lines
  • Our most characterized serum with the lowest endotoxin and hemoglobin levels
  • Endotoxin level: ≤5 EU/mL
  • Hemoglobin level: ≤20 mg/dL
  • Origin: Refer to CoA for specific origin (US, Australia, New Zealand)
  • US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US
Gibco Serum Delivers
  • ISO 13485-certified; processed in FDA-registered facilities
  • Triple filtered at 0.1 μm
  • Gibco bottle easier to use in hood, reduces risk of contamination and helps improve consistency
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook