Use promo code "25339" to get your promotional price.
Sartorius™ Cubis™ II Semi-micro MCA 226S Balance with EC Type Approval Certificate (EU)
The Cubis™ II range of semi-micro balances with a maximum load between 120 g and 220 g and a readability between 0.005 mg and 0.01 mg provide the ideal choice for a broad range of applications.
28445.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
Additional Details:
Weight:
14.30000kg
Specification
Certifications/Compliance | EC Type Approval Certificate (for EU except France) |
---|---|
Capacity | 220 g |
Display Type | Color TFT Touchscreen |
Readability | 0.005 mg |
Model | QP99 HWL Configuration |
Stabilization Time | 1.5 s |
Type | Semi-micro Balance |
Width (Metric) | 240 mm |
Height (Metric) | 301 mm |