Invitrogen™ Microplates for Fluorescence-based Assays
Microplates for fluorescence-based assays are 96- and 384-well, black-walled, clear-bottom microplates that are optimized for fluorescence-based applications on standard fluorescence microplate readers.
Additional Details:
Weight:
1.95000kg
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specification
| Barcode | No Barcode |
|---|---|
| Format | Standard |
| Content And Storage | Store at room temperature. |
| Array | 8 x 12 |
| Autoclavable | Not Autoclavable |
| Packaging | 30-plates |
| Product Line | Invitrogen |
| Color | Clear Bottom,Black |
| Color | Clear Bottom,Black |
| Disposable | Single-use |
| Product Type | Microplates |
| Material | Polystyrene |
| No. of Wells | 96 |
| Volume (Metric) Working | 200 μL |
| Sterility | Non-sterile |
| Well Shape | Flat Bottom (F-well) |
| For Use With (Application) | Nucleic Acid Quantification |
| Quantity | 30 Plates |