Learn More
Waters Corporation Waters PAH C18 Column
Polyaromatic hydrocarbons (PAHs) are among the most frequently monitored environmental contaminants.
Specifications
Max. Pressure | 6000 psi (415 Bar) |
---|---|
Particle Shape | Spherical |
Pore Size | 120 Å |
Quantity | 1/Ea. |
USP Type | L1 |
Description
- Standard and official methods for the analysis of PAHs are found in compendia for air, drinking water, wastewater, solid waste, and food analysis.
- Many of these methods specify HPLC, usually with UV and fluorescence detection, as the recommended analytical procedure.
- Introducing, from Waters Corporation, world leaders in HPLC and LCMS, the best column for HPLC analysis of PAHs, Waters PAH.
Specifications
6000 psi (415 Bar) | |
120 Å | |
L1 | |
Column |
Spherical | |
1/Ea. | |
Yes | |
Polyaromatic Hydrocarbon |