78.29 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
Color | White |
---|---|
Material | Polypropylene |
No. per Case | 200 Cs. |
Liner Material | Foam/LDPE |
Neck Finish | 28-410 |
Certifications/Compliance | ISO 9001 |
For Use With (Equipment) | Glass bottle or vial |