DWK Life Sciences Wheaton™ White Polypropylene Caps with Foam/LDPE Liners

For general storage and packaging applications

Additional Details:
Additional Details: Weight: 0.45360kg



Product Code. 12498762

Quantity Price
1 64.45  € / Pack of 200
Estimated Shipment
31-10-2025
Add to basket


  • Description and Specification
  • Alternative Products
Description and Specification

Specification

Color White
Material Polypropylene
No. per Case 200 Cs.
Liner Material Polyethylene Foam
Neck Finish 20-410
Certifications/Compliance ISO 9001
For Use With (Equipment) Glass bottle or vial

  • Lined with foam polyethylene
  • Excellent barrier to aqueous solutions
  • Repeat sealability

Alternative Products