Learn More
Fisherbrand™ White Polypropylene Caps
Fisherbrand™ White Polypropylene Caps include a clear writing surface for easy identification.
93.00 € - 422.00 €
Product Code | Brand | Closure Size | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Size | Quantity | Price | Quantity & Availability | ||||
16144617
|
Fisherbrand™
FB02912102A |
8-425 mm | 500 |
164.00 €
Pack of 500 |
Estimated Shipment: 22-10-2025
Log in to see stock. |
||||
16114597
|
Fisherbrand™
FB02912064A |
13-425 mm | 500 |
190.00 €
Pack of 500 |
Estimated Shipment: 18-08-2025
Log in to see stock. |
||||
16104647
|
Fisherbrand™
FB02912291A |
48-400 mm | 576 |
422.00 €
Pack of 576 |
Estimated Shipment: 22-10-2025
Log in to see stock. |
||||
16144657
|
Fisherbrand™
FB02912332A |
48-400 mm | 1700 |
261.00 €
Pack of 1700 |
Estimated Shipment: 22-10-2025
Log in to see stock. |
||||
16194607
|
Fisherbrand™
FB02912088A |
70-400 mm | 144 |
93.00 €
Pack of 144 |
Estimated Shipment: 22-10-2025
Log in to see stock. |
||||
Description
- Foamed polyethylene liner provides tight seal and chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
- PTFE faced foamed polyethylene liner offers chemical resistance excellent for environmental samples, pharmaceuticals and diagnostic reagents
- Poly-vinyl liner provides an excellent moisture barrier good for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Specifications
White | |
Polypropylene |