Learn More
YAP1 Rabbit anti-Human, Mouse, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13584-1-AP-150UL
 View more versions of this product
Product Code. 16811874
  • 410.00 € / 150µL
Description
This antibody is a rabbit polyclonal antibody. The calculated molecular weight of YAP1 is 54 kDa, but phosphorylated YAP1 is about 65 kDa.
YAP1 (Yes-Associated Protein) is a founding member of the WW domain family of proteins, transcriptional co-activator and oncogene. Its gene is transcriptionally activated in squamous cell carcinoma cells. YAP can act both as a coactivator and corepressor and is the critical downstream regulatory target in the Hippo signaling pathway. The Hippo signaling pathway plays a pivotal role in organ size control and tumor suppression by restricting proliferation and promoting apoptosis. Also plays key roles in tissue tension, 3D tissue shape, cell proliferation, cell death, and cell migration. Mutations in the IRF5 gene results in coloboma.
Specifications
Specifications
YAP1
Polyclonal
Unconjugated
YAP1
65 kDa Yes associated protein, YAP, YAP1, YAP2, YAP65, YKI, Yorkie homolog
Rabbit
Antigen Affinity Chromatography
RUO
10413, 22601, 561411
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P46937, P46938, Q1L8J7
yap1
YAP1 Fusion Protein Ag4510
150 μL
Primary
Human, Mouse, Zebrafish