Memmert™ 4 - 20 mA Current Loop Interface

4-20mA current loop interface temperature controller

Additional Details:
Additional Details: Weight: 0.01100kg



Product Code. 17110145

Quantity Price
1 242.0  € / Each
Estimated Shipment
08-10-2025
Add to basket


Description and Specification

Specification

For Use With (Equipment) IPP1400eco
Type Temperature Controller

  • Temperature controller
  • Actual value; -10 to +80 °C = 4 -20 mA