Learn More
Memmert™ 4 - 20 mA Current Loop Interface
4-20mA current loop interface temperature controller
242.00 €
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
17109734
|
Memmert™
HPP1400ecoV3 |
HPP1400eco |
242.00 €
Each |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
17110125
|
Memmert™
IPP110ecoV3 |
IPP110eco |
242.00 €
Each |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
17110145
|
Memmert™
IPP1400ecoV3 |
IPP1400eco |
242.00 €
Each |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
17120165
|
Memmert™
IPP2200ecoV3 |
IPP2200eco |
242.00 €
Each |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
17129804
|
Memmert™
HPP260ecoV3 |
HPP260eco |
242.00 €
Each |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
17129894
|
Memmert™
HPP750ecoV3 |
HPP750eco |
242.00 €
Each |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
17130205
|
Memmert™
IPP260ecoV3 |
IPP260eco |
242.00 €
Each |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
17130245
|
Memmert™
IPP410ecoV3 |
IPP410eco |
242.00 €
Each |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
17149754
|
Memmert™
HPP2200ecoV3 |
HPP2200eco |
242.00 €
Each |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
17159704
|
Memmert™
HPP110ecoV3 |
HPP110eco |
242.00 €
Each |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
17160075
|
Memmert™
IPP1060ecoV3 |
IPP1060eco |
242.00 €
Each |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
17160285
|
Memmert™
IPP750ecoV3 |
IPP750eco |
242.00 €
Each |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
17169844
|
Memmert™
HPP410ecoV3 |
HPP410eco |
242.00 €
Each |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
17189654
|
Memmert™
HPP1060ecoV3 |
HPP1060eco |
242.00 €
Each |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
Description
- Temperature controller
- Actual value; -10 to +80 °C = 4 -20 mA
Specifications
Temperature Controller |