Code:
Z1
Greiner Bio-One CELLSTAR 96-well, Cell Culture-Treated, Flat-Bottom Microplate
Color: Clear; Sterility: Sterile; Lid: Without Lid
Additional Details:
Weight:
5.80000kg
Specification
Certifications/Compliance | Free of detectable DNase/RNase, human DNA and pyrogens; contents non-cytotoxic |
---|---|
Format | Standard |
Lid | Without Lid |
Culture Area | 34 mm2 |
Array | 8 x 12 |
Autoclavable | Not Autoclavable |
Packaging | 5/Pk., 100/Cs. |
Color | Clear |
Material | Polystyrene |
No. of Wells | 96 |
Volume (Metric) Working | 50 to 335 μL |
Sterility | Sterile |
Surface Treatment | Cell Culture |
Well Shape | Flat Bottom (F-well) |
Max. RCF | 4800 x g (swinging bucket rotor) |
No. per Case | 100 |
No. per Pack | 5 |
Quantity | 100/Cs. |
Temperature Range (Metric) | -20°C to +60°C |
Well Design | Chimney |
Dimensions (L x W x H) | 128 x 85 x 15 mm |