Gibco™ N-2 Supplement (100X)
N-2 Supplement is a versatile serum-free cell culture supplement that supports the growth and viability of various cell types.
Overview
760.03 GBP valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Code: NEW
Additional Details:
Additional Details: CAS Number: 10102-18-8 Weight: 0.05000kg
Disclaimers: For Research Use Only. Not for use in diagnostic procedures.
Product Code. 15410294
Quantity Price
1 £ 1140.0 / 50mL
In stock, ready to ship: 675
Add to basket
  • Description and Specification
  • Alternative Products
  • Safety and Handling
  • Special Offers
Description and Specification
Specification
Content And Storage Store in freezer (-5 to -20°C) and protect from light.
Cell Type Neural Cells
Classification Serum-free
Concentration 100 X
Culture Type Mammalian Cell Culture
Form Liquid
Product Type Cell Culture Supplement
Serum Level Serum-Free
Sterility Sterile
pH 6 to 8
Quantity 50 mL
Shipping Condition Dry Ice
Source Serum-Free
Tested For Endotoxin, Performance, Sterility
N-2 Supplement has been demonstrated in wide-ranging applications, such as the culture of neural stem cells, the culture of the SH-SY5Y cell line (subline of the neuroblastoma cell line SK-N-SH), and culture of the HT22 mouse hippocampal neuronal cell line. Additionally, N-2 Supplement adds support to cell culture media used to differentiate pluripotent stem cells (PSCs) to a variety of cell types, including neural and pancreatic-like cell types.
N-2 Supplement may be used with Neurobasal media supplemented with growth factors such as bFGF and EGF. When combined with Dulbecco's Modified Eagle Medium (D-MEM) and fetal bovine serum (FBS), N-2 Supplement also provides suitable supplementation for the culture of human and rodent astrocytes.
  • Each lot is cGMP manufactured
  • Each lot is tested for sterility, endotoxin, and performance
  • Contains insulin, iron-rich holo-transferrin, progesterone, putrescine, and selenite
  • Can be used as a substitute for Bottenstein's N-1 formulation
  • Provided as a 100X concentrate
Choose Gibco N-2 Supplement for a reliable, high-quality product trusted by researchers worldwide.
Alternative Products
Safety and Handling
[Mercuric acetate]
[Danger]
[{code=ACUTE TOX. 1, sequence=1, description=Acute toxicity Category 1}, {code=ACUTE TOX. 2, sequence=2, description=Acute toxicity Category 2}, {code=AQUA CHRONIC 1, sequence=3, description=LONG-TERM AQUATIC HAZARD Chronic 1}, {code=AQUATIC ACUTE 1, sequence=4, description=ACUTE AQUATIC Acute 1}, {code=STOT RE 2, sequence=5, description=Specific target organ toxicity after repeated exposure Category 2}]
[{code=H300, sequence=1, description=Fatal if swallowed.}, {code=H310, sequence=2, description=Fatal in contact with skin.}, {code=H330, sequence=3, description=Fatal if inhaled.}, {code=H373, sequence=4, description=May cause damage to organs.}, {code=H400, sequence=5, description=Very toxic to aquatic life.}, {code=H410, sequence=6, description=Very toxic to aquatic life with long lasting effects.}]
[{code=P260, sequence=51, description=Do not breathe dust/fume/gas/mist/vapours/spray.}, {code=P273, sequence=52, description=Avoid release to the environment.}, {code=P280, sequence=53, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P302+P352, sequence=54, description=IF ON SKIN: Wash with plenty of water/soap.}, {code=P304+P340, sequence=55, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P310, sequence=56, description=Immediately call a POISON CENTER/doctor/.}, {code=P362, sequence=57, description=Take off contaminated clothing.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : Mercuric acetate}]
[{code=GHS06, sequence=1, description=SKULL & CROSSBONES}, {code=GHS08, sequence=2, description=HEALTH HAZARD}, {code=GHS09, sequence=3, description=ENVIRONMENT}]
Product Identifier
  • Mercuric acetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H300- Fatal if swallowed.
  • H310- Fatal in contact with skin.
  • H330- Fatal if inhaled.
  • H373- May cause damage to organs.
  • H400- Very toxic to aquatic life.
  • H410- Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260- Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273- Avoid release to the environment.
  • P280- Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352- IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310- Immediately call a POISON CENTER/doctor/.
  • P362- Take off contaminated clothing.
Supplemental Information
  • MIXTURE LIST- Contains : Mercuric acetate
Special Offers