Honeywell Research Chemicals Chlorobenzene, ReagentPlus™, 99%, Honeywell™



Quantity | 1 L |
---|---|
Packaging | Glass Bottle |
Chemical Identifiers
CAS | 108-90-7 |
---|---|
Molecular Formula | C6H5Cl |
Molecular Weight (g/mol) | 112.556 |
MDL Number | MFCD00000530 |
InChI Key | MVPPADPHJFYWMZ-UHFFFAOYSA-N |
Synonym | monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene |
PubChem CID | 7964 |
ChEBI | CHEBI:28097 |
IUPAC Name | chlorobenzene |
SMILES | C1=CC=C(C=C1)Cl |
Specification
Chemical Name or Material | Chlorobenzene |
---|---|
Melting Point | -45°C |
CAS | 108-90-7 |
Color | Colorless |
Density | 1.106g/mL at 25°C |
Boiling Point | 130°C to 132°C |
Quantity | 1 L |
Explosion Limit | 0.071 |
Flash Point | 27°C (80.6°F) |
Ignition Point | 1178°F |
Packaging | Glass Bottle |
Physical Form | Liquid |
Refractive Index | n20/D 1.524(lit.) |
Molecular Formula | C6H5Cl |
MDL Number | MFCD00000530 |
Vapor Pressure | 11.8mmHg (25°C) |
UN Number | UN1134 |
Beilstein | 605632 |
Synonym | monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene |
Synonym | monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene |
Synonym | monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene |
Synonym | monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene |
Synonym | monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene |
Synonym | monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene |
Synonym | monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene |
Synonym | monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene |
Synonym | monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene |
Synonym | monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene |
Solubility Information | Soluble in water, 0.207g/L at 20°C |
InChI Key | MVPPADPHJFYWMZ-UHFFFAOYSA-N |
SMILES | C1=CC=C(C=C1)Cl |
IUPAC Name | chlorobenzene |
Molecular Weight (g/mol) | 112.556 |
PubChem CID | 7964 |
ChEBI | CHEBI:28097 |
Formula Weight | 112.56g/mol |
Percent Purity | 99% |
Grade | ReagentPlus™ |
Technical documents
[Chlorobenzene]
[Warning]
[{code=FLAM. LIQ. 3, sequence=1, description=Flammable liquid Category 3}, {code=ACUTE TOX. 4, sequence=2, description=Acute toxicity Category 4}, {code=SKIN IRRIT. 2, sequence=3, description=Skin corrosion/irritation Category 2}, {code=AQUA CHRONIC 2, sequence=4, description=LONG-TERM AQUATIC HAZARD Chronic 2}]
[{code=H226, sequence=1, description=Flammable liquid and vapour.}, {code=H315, sequence=2, description=Causes skin irritation.}, {code=H332, sequence=3, description=Harmful if inhaled.}, {code=H411, sequence=4, description=Toxic to aquatic life with long lasting effects.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P260, sequence=52, description=Do not breathe dust/fume/gas/mist/vapours/spray.}, {code=P280, sequence=53, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P284, sequence=54, description=¢In case of inadequate ventilation! wear respiratory protection.}, {code=P302+P352, sequence=55, description=IF ON SKIN: Wash with plenty of water/.}, {code=P304+P340, sequence=56, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P308+P313, sequence=57, description=IF exposed or concerned: Get medical advice/attention.}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}, {code=GHS09, sequence=3, description=ENVIRONMENT}]
Product Identifier
- Chlorobenzene
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H226- Flammable liquid and vapour.
- H315- Causes skin irritation.
- H332- Harmful if inhaled.
- H411- Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260- Do not breathe dust/fume/gas/mist/vapours/spray.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P284- ¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352- IF ON SKIN: Wash with plenty of water/.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313- IF exposed or concerned: Get medical advice/attention.