missing translation for 'onlineSavingsMsg'
Learn More

Chlorobenzene, ReagentPlus™, 99%, Honeywell™

Reagent Grade, 99%

42.71 € - 431.85 €

Chemical Identifiers

CAS 108-90-7
Molecular Formula C6H5Cl
Molecular Weight (g/mol) 112.556
MDL Number MFCD00000530
InChI Key MVPPADPHJFYWMZ-UHFFFAOYSA-N
Synonym monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene
PubChem CID 7964
ChEBI CHEBI:28097
IUPAC Name chlorobenzene
SMILES C1=CC=C(C=C1)Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15600950
SDS View Documents
Honeywell Research Chemicals
101389-1L
1 L Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15610950
SDS View Documents
Honeywell Research Chemicals
101389-2.5L
2.5 L Glass Bottle
77.37 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15620950
SDS View Documents
Honeywell Research Chemicals
101389-20L
20 L Stainless Steel Can This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

108-90-7
112.556
MVPPADPHJFYWMZ-UHFFFAOYSA-N
7964
chlorobenzene
C6H5Cl
MFCD00000530
monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene
CHEBI:28097
C1=CC=C(C=C1)Cl

Specifications

-45°C
Colorless
130°C to 132°C
27°C (80.6°F)
Liquid
C6H5Cl
11.8mmHg (25°C)
605632
Soluble in water, 0.207g/L at 20°C
C1=CC=C(C=C1)Cl
112.556
CHEBI:28097
99%
Chlorobenzene
108-90-7
1.106g/mL at 25°C
0.071
1178°F
n20/D 1.524(lit.)
MFCD00000530
UN1134
monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene
MVPPADPHJFYWMZ-UHFFFAOYSA-N
chlorobenzene
7964
112.56g/mol
ReagentPlus™
Videos
Safety and Handling

Safety and Handling

P210-P261-P370 + P378

H226-H315-H332-H411

missing translation for 'dotInformation' : class 3 - PG 3 - Chlorobenzene

missing translation for 'einecsNumber' : 203-628-5

missing translation for 'rtecsNumber' : CZ0175000

Product Identifier
  • Chlorobenzene

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H332-Harmful if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.