Learn More
Buerkle™ LPDE Wide Mouth Wash Bottles
Manufactured from LDPE for strength, clarity, and flexibility. Buerkle™ LDPE Wide Mouth Wash Bottles are imprinted with substance name in German, English, French and Spanish, hazard and safety instructions, GHS pictograms and CAS number.
Brand: Buerkle™ 0310-3055
10.65 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Wide mouth opening for safe filling
- Yellow translucent color with light protection
- Curved aperture on screw cap for precision jet dispensing without splatters
- Color coded closures
Specifications
500 mL | |
39 mm | |
Red | |
Wide | |
Acetone |
Translucent Yellow | |
LDPE | |
Yes | |
Wash Bottle |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.