missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ LPDE Wide Mouth Wash Bottles
Manufactured from LDPE for strength, clarity, and flexibility. Buerkle™ LDPE Wide Mouth Wash Bottles are imprinted with substance name in German, English, French and Spanish, hazard and safety instructions, GHS pictograms and CAS number.
11.83 € - 12.10 €
Specifications
Capacity (Metric) | 500 mL |
---|---|
Color | Translucent Yellow |
Diameter (Metric) Inner Neck | 39 mm |
Material | LDPE |
Closure Included | Yes |
Product Code | Brand | Closure Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Color | Price | Quantity & Availability | |||||
10325764
|
Buerkle™
0310-3054 |
Blue |
11.83 €
Each |
In Stock
Log in to see stock. |
|||||
10775685
|
Buerkle™
0310-3052 |
Green |
11.83 €
Each |
In Stock
Log in to see stock. |
|||||
10448331
|
Buerkle™
0310-3053 |
Orange |
11.83 €
Each |
In Stock
Log in to see stock. |
|||||
10765685
|
Buerkle™
0310-3055 |
Red |
11.83 €
Each |
In Stock
Log in to see stock. |
|||||
10336194
|
Buerkle™
0310-3051 |
Transparent |
11.83 €
Each |
In Stock
Log in to see stock. |
|||||
10142644
|
Buerkle™
0310-3050 |
Yellow |
12.10 €
Each |
In Stock
Log in to see stock. |
|||||
Description
- Wide mouth opening for safe filling
- Yellow translucent color with light protection
- Curved aperture on screw cap for precision jet dispensing without splatters
- Color coded closures
Specifications
500 mL | |
39 mm | |
Yes | |
Wash Bottle |
Translucent Yellow | |
LDPE | |
Wide |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title