BRAND™ Robotic Tips, BIO-CERT™ Liquid Handling, Sterile
Pipette tips offer reliable quality for precise and demanding analyses with the pipetting robot Liquid Handling Station.
Overview
101.85 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Additional Details:
Additional Details: Weight: 0.53330kg
Product Code. 17506696
Quantity Price
1 130.0  € / Pack of 960
Estimated Shipment
08-08-2025
Add to basket
Description and Specification
Specification
Volume (Metric) 1 to 50 μL
Filtered Filtered
Sterility Sterile
Packaging TipRack (960/Pk.)
Product Type Robotic Tips
Material Polypropylene
  • The raw materials are free from DiHEMDA und Oleamid. Tips are manufactured at BRAND under monitored cleanroom conditions and are automatically packaged. No parting agents or demolding aids are used. All racked tips and filter tips up to 1000 μL are free of DNA (< 40 fg), RNase (< 8.6 fg), endotoxins (< 1 pg) and ATP (< 1 fg).
  • For use with pipetting robots, tips are required that are matched to the robot's mode of operation in terms of geometry and surface properties. BRAND robotic pipette tips and robotic filter tips meet all requirements for use with the Liquid Handling Station pipetting robot.
Special Offers