Semperit™ Semperguard™ Green Nitrile Disposable Gloves

Developed with sustainability in mind due to the unique production process, the glove offers skin-friendly hand protection, reduces the risk of allergy and feels soft to the skin. Comfortable and strong tensile strength ensures stability and a pleasant wearing experience even over longer periods.

Additional Details:
Additional Details: Weight: 0.77400kg



Product Code. 15970057

 N/A
N/A

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


  • Description and Specification
  • Alternative Products
Description and Specification

Specification

Certifications/Compliance (Europe) PPE Category III, EN 420, EN ISO 374-1, EN 374-2, EN 16523-1, EN 374-4, EN ISO 374-5, EN 455-1, EN 455-2, EN 455-3, EN 455-4, ISO 2859-1, ASTM D6319, ASTM F1671
Chlorinated No
Accelerator Free Yes
Applications Industrial
AQL 1.5
Color Mint Green
Disposable Single-use
Exterior Finish Fingertip Textured
Material Nitrile
Powdered Powder Free
Size X-Large
Sterility Non-sterile
Glove Cuff Style Rolled
Series Semperguard™ Green
Tensile Strength 6 Newton
Length (Metric) 24 cm

  • PPE Cat. III
  • AQL 1.5
  • Powder-free
  • Dispoable, non-sterile
  • Comfortable to wear, even over longer periods of time
  • Skin-friendly formulation: no chlorine, no natural rubber latex proteins, no rubber accelerators
  • Manufactured while saving on water and energy, and reducing CO2 emissions
  • Synthetic inner coating
  • Textured fingertips provide extra grip
  • Stability and pleasant wearing experience
  • Glove material is resistant to blood and other body fluids, and protects against bacteria, fungi and viruses including SARS-CoV-2, Hepatitis B, Hepatitis C, and HIV
  • Ideal for applications including Personal Protection, Food/Catering, Consumer/Retail, Beauty/Body Art and Hairdressing

Alternative Products