missing translation for 'onlineSavingsMsg'
Learn More

Semperit™ Semperguard™ Green Nitrile Disposable Gloves
GREENER_CHOICE

Developed with sustainability in mind due to the unique production process, the glove offers skin-friendly hand protection, reduces the risk of allergy and feels soft to the skin. Comfortable and strong tensile strength ensures stability and a pleasant wearing experience even over longer periods.

33.00 € - 35.00 €

Specifications

Certifications/Compliance (Europe) PPE Category III, EN 420, EN ISO 374-1, EN 374-2, EN 16523-1, EN 374-4, EN ISO 374-5, EN 455-1, EN 455-2, EN 455-3, EN 455-4, ISO 2859-1, ASTM D6319, ASTM F1671
Chlorinated No
Accelerator Free Yes
Applications Industrial
AQL 1.5
View More Specs

Products 5
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15930057
GREENER_CHOICE
View Documents
Semperit™
3000008212
X-Small
34.00 €
Pack of 200
In Stock
Add to basket
 
15940057
GREENER_CHOICE
View Documents
Semperit™
3000008213
Small This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15950057
GREENER_CHOICE
View Documents
Semperit™
3000008214
Medium
35.00 €
Pack of 200
In Stock
Add to basket
 
15960057
GREENER_CHOICE
View Documents
Semperit™
3000008215
Large This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15970057
GREENER_CHOICE
View Documents
Semperit™
3000008216
X-Large This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • PPE Cat. III
  • AQL 1.5
  • Powder-free
  • Dispoable, non-sterile
  • Comfortable to wear, even over longer periods of time
  • Skin-friendly formulation: no chlorine, no natural rubber latex proteins, no rubber accelerators
  • Manufactured while saving on water and energy, and reducing CO2 emissions
  • Synthetic inner coating
  • Textured fingertips provide extra grip
  • Stability and pleasant wearing experience
  • Glove material is resistant to blood and other body fluids, and protects against bacteria, fungi and viruses including SARS-CoV-2, Hepatitis B, Hepatitis C, and HIV
  • Ideal for applications including Personal Protection, Food/Catering, Consumer/Retail, Beauty/Body Art and Hairdressing
Specifications

Specifications

PPE Category III, EN 420, EN ISO 374-1, EN 374-2, EN 16523-1, EN 374-4, EN ISO 374-5, EN 455-1, EN 455-2, EN 455-3, EN 455-4, ISO 2859-1, ASTM D6319, ASTM F1671
Yes
1.5
Single-use
Nitrile
Non-sterile
Semperguard™ Green
24 cm
No
Industrial
Mint Green
Fingertip Textured
Powder Free
Rolled
6 Newton
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.