Learn More
Ohaus™ Stainless Steel Calibration Weights - OIML E2, NVLAP
Supplied with a NVLAP Accredited Certificate and Adjusted to Meet or Exceed OIML E2 Tolerances
80.15 € - 2400.00 €
Specifications
Certifications/Compliance | NVLAP Accredited Certificate |
---|---|
Includes | Protective Case |
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
16632382
|
Ohaus™
80780284 |
1 mg |
80.15 €
Each |
In Stock
Log in to see stock. |
|||||
16642382
|
Ohaus™
80780285 |
2 mg |
80.15 €
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
16652382
|
Ohaus™
80780286 |
5 mg |
80.15 €
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
16662382
|
Ohaus™
80780287 |
10 mg |
80.15 €
Each |
In Stock
Log in to see stock. |
|||||
16672382
|
Ohaus™
80780288 |
20 mg |
80.15 €
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
16682382
|
Ohaus™
80780289 |
50 mg |
80.15 €
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
16692382
|
Ohaus™
80780290 |
100 mg |
80.15 €
Each |
In Stock
Log in to see stock. |
|||||
16602392
|
Ohaus™
80780291 |
200 mg |
80.15 €
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
16612392
|
Ohaus™
80780292 |
500 mg |
80.15 €
Each |
In Stock
Log in to see stock. |
|||||
16622392
|
Ohaus™
80780293 |
1 g |
124.00 €
Each |
In Stock
Log in to see stock. |
|||||
16632392
|
Ohaus™
80780294 |
2 g |
129.00 €
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
16642392
|
Ohaus™
80780295 |
5 g |
135.00 €
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
16652392
|
Ohaus™
80780296 |
10 g |
140.00 €
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
16662392
|
Ohaus™
80780297 |
20 g |
151.00 €
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
16672392
|
Ohaus™
80780298 |
50 g |
166.00 €
Each |
In Stock
Log in to see stock. |
|||||
16682392
|
Ohaus™
80780299 |
100 g |
178.00 €
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
16692392
|
Ohaus™
80780300 |
200 g |
222.00 €
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
16602402
|
Ohaus™
80780301 |
500 g |
333.00 €
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
16612402
|
Ohaus™
80780302 |
1 kg |
460.00 €
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
16622402
|
Ohaus™
80780303 |
2 kg |
708.00 €
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
16632402
|
Ohaus™
80780304 |
5 kg |
979.00 €
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
16642402
|
Ohaus™
80780305 |
10 kg |
1465.00 €
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
16652402
|
Ohaus™
80780306 |
20 kg |
2400.00 €
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
Description
- Weights in masses ranging from 1 gram to 20 kilograms are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free
- NVLAP Calibration certificate is accredited to ISO 17025 which is recognizable through ILAC
- Weight surfaces meet or exceed OIML R 111 and have a highly polished, mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifications
NVLAP Accredited Certificate | |
Protective Case |