Learn More
Falcon™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate
Color: Clear; Sterility: Sterile; Lid: With Lid
Brand: Falcon™ 353936
342.76 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Includes: Lids
Description
Available with tissue culture-treated and nontreated surfaces. Made of crystal grade virgin polystyrene for optimum visibility.
- Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
- Opaque walls prevent well-to-well cross-talk.
- Optically clear flat bottom permits direct microscopic viewing.
- Crystal-grade virgin polystyrene
- Sterilized by gamma irradiation
- Nonpyrogenic
- Non-treated polystyrene, standard tissue culture (TC), and Corning™ PrimariaTM surface treatments available
- Convenient, peel-open medical-style packaging for individual product presentation
- Individual and Ready-Stack (RS) trays are PET (Code 1) and recyclable
- Falcon 96, 384, and 1536 well microplates are compatible with automation systems and meet standard ANSI/SBS footprint dimensions for microplates
Order Info
Packaged sterile, 14 plates per tray
Specifications
Standard | |
0.32 cm2 | |
TC-Treated | |
Cell Culture Plate | |
96 | |
Sterile | |
0.37 mL / 370 μL | |
84/Cs. | |
84/Cs. |
With Lid | |
8 x 12 | |
Clear | |
Polystyrene | |
40−275 μL | |
Cell Culture | |
Flat Bottom (F-well) | |
14/Pk. |